MilliporeSigma
  • PI3K/Akt signaling pathway may be involved in MCP-1-induced P2X4R expression in cultured microglia and cancer-induced bone pain rats.
PI3K/Akt signaling pathway may be involved in MCP-1-induced P2X4R expression in cultured microglia and cancer-induced bone pain rats.
Neuroscience letters (2019-02-19)
Ying Yan, Yongxin Liang, Tao Ding, Haichen Chu
ABSTRACT
P2X4 receptor (P2X4R), a subtype of P2 purinergic receptors, is an ATP-gated receptor through which activity of spinal microglia instigates pain hypersensitivity in various pain conditions. Accumulating evidence indicates that monocyte chemoattractant protein-1 (MCP-1) plays an important role in chronic pain facilitation, and it could stimulate microglia activation and involve in regulating P2X4R expression. However, the mechanism of MCP-1 in regulating the expression of P2X4R in microglia is poorly understood, and whether MCP-1 can aggravate pain via up-regulating spinal P2X4R expression in Cancer-induced Bone Pain (CIBP) remains unclear. In this study, we observed that Iba-1 and P2X4R expression is increased in microglia treated with MCP-1, and blockade with a selective CCR2 antagonist RS-504393 suppressed microglia activation and reduced P2X4R expression in cultured microglia. In response to MCP-1, the expression level of p-Akt was also increased and RS-504393 inhibited the increase. Besides, PI3K inhibitor LY 294002 could attenuate MCP-1-induced P2X4R expression in cultured microglia. MCP-1 was found to be associated with P2X4R expression and mechanical allodynia induced by CIBP in vivo since the expression of MCP-1 was increased in CIBP and RS-504393 alleviated the P2X4R expression and mechanical allodynia in CIBP. Moreover, RS-504393 also reduced the increase of p-Akt induced by CIBP. Inhibition of PI3K/Akt pathway may partly reduce MCP-1/CCR2-induced expression of P2X4R and mechanical allodynia in CIBP rats.