MilliporeSigma
  • Orthonasal and retronasal detection thresholds of 26 aroma compounds in a model alcohol-free beer: Effect of threshold calculation method.
Orthonasal and retronasal detection thresholds of 26 aroma compounds in a model alcohol-free beer: Effect of threshold calculation method.
Food research international (Ottawa, Ont.) (2019-07-10)
José A Piornos, Alexia Delgado, Rémi C J de La Burgade, Lisa Methven, Dimitrios P Balagiannis, Elisabeth Koussissi, Eric Brouwer, Jane K Parker
ABSTRACT
Detection thresholds are used routinely to determine the odour-active compounds in foods. The composition of a food matrix, such as hydrophobicity or solids content, has an impact on the release of flavour compounds, and thus on thresholds. In the case of beer, thresholds determined in alcoholic beer may not be the same for alcohol-free beer (AFB). Therefore, the aim of this study was to determine detection thresholds for aroma compounds typically found in beer, within a model AFB. The model was designed to match the sugar concentration and pH of an AFB brewed by a cold contact process. Thresholds were measured using a 3-AFC procedure and calculated using either Best Estimate Threshold (BET) method or by logistic regression. Moreover, an algorithm for the removal of false positives was applied to adjust the assessors' raw responses. Retronasal thresholds were generally lower than orthonasal. Those calculated by BET were significantly higher (p < 0.05) than those from logistic regression, and removal of false positives also produced significantly higher thresholds than those from raw data. The use of logistic regression has the advantage of providing the mathematical model describing the behaviour of the group. The results from this study can be used to better understand the role of flavour compounds in AFB and the effect of the calculation method to prevent under- or overestimated results.