MilliporeSigma
  • Targeted genetic screening in mice through haploid embryonic stem cells identifies critical genes in bone development.
Targeted genetic screening in mice through haploid embryonic stem cells identifies critical genes in bone development.
PLoS biology (2019-07-03)
Meizhu Bai, Yujiao Han, Yuxuan Wu, Jiaoyang Liao, Lin Li, Lijun Wang, Qing Li, Wenhui Xing, Luonan Chen, Weiguo Zou, Jinsong Li
ABSTRACT
Mutagenic screening is powerful for identifying key genes involved in developmental processes. However, such screens are successful only in lower organisms. Here, we develop a targeted genetic screening approach in mice through combining androgenetic haploid embryonic stem cells (AG-haESCs) and clustered regularly interspaced palindromic repeats/CRISPR-associated protein 9 (CRISPR-Cas9) technology. We produced a mutant semi-cloned (SC) mice pool by oocyte injection of AG-haESCs carrying constitutively expressed Cas9 and an single guide RNA (sgRNA) library targeting 72 preselected genes in one step and screened for bone-development-related genes through skeletal analysis at birth. This yielded 4 genes: Zic1 and Clec11a, which are required for bone development, and Rln1 and Irx5, which had not been previously considered. Whereas Rln1-/- mice exhibited small skeletal size only at birth, Irx5-/- mice showed skeletal abnormalities both in postnatal and adult phases due to decreased bone mass and increased bone marrow adipogenesis. Mechanistically, iroquois homeobox 5 (IRX5) promotes osteoblastogenesis and inhibits adipogenesis by suppressing peroxisome proliferator activated receptor γ (PPARγ) activation. Thus, AG-haESC-mediated functional mutagenic screening opens new avenues for genetic interrogation of developmental processes in mice.
Product Description
SAFC
Insulin, Human Recombinant, for research or for further manufacturing use, dry powder