MilliporeSigma
  • Hydrogen Sulfide as a Toxic Product in the Small-Large Intestine Axis and its Role in IBD Development.
Hydrogen Sulfide as a Toxic Product in the Small-Large Intestine Axis and its Role in IBD Development.
Journal of clinical medicine (2019-07-25)
Ivan Kushkevych, Dani Dordević, Peter Kollar, Monika Vítězová, Lorenzo Drago
ABSTRACT
The small-large intestine axis in hydrogen sulfide accumulation and testing of sulfate and lactate in the gut-gut axis of the intestinal environment has not been well described. Sulfate reducing bacteria (SRB) of the Desulfovibrio genus reduce sulfate to hydrogen sulfide and can be involved in ulcerative colitis development. The background of the research was to find correlations between hydrogen sulfide production under the effect of an electron acceptor (sulfate) and donor (lactate) at different concentrations and Desulfovibrio piger Vib-7 growth, as well as their dissimilatory sulfate reduction in the intestinal small-large intestinal environment. Microbiological, biochemical, and biophysical methods, and statistical processing of the results (principal component and cross-correlation analyses) were used. D. piger Vib-7 showed increased intensity of bacterial growth and hydrogen sulfide production under the following concentrations of sulfate and lactate: 17.4 mM and 35.6 mM, respectively. The study showed in what kind of intestinal environment D. piger Vib-7 grows at the highest level and produces the highest amount of hydrogen sulfide. The optimum intestinal environment of D. piger Vib-7 can serve as a good indicator of the occurrence of inflammatory bowel diseases; meaning that these findings can be broadly used in medicine practice dealing with the monitoring and diagnosis of intestinal ailments.