MilliporeSigma
  • An aptasensor for troponin I based on the aggregation-induced electrochemiluminescence of nanoparticles prepared from a cyclometallated iridium(III) complex and poly(4-vinylpyridine-co-styrene) deposited on nitrogen-doped graphene.
An aptasensor for troponin I based on the aggregation-induced electrochemiluminescence of nanoparticles prepared from a cyclometallated iridium(III) complex and poly(4-vinylpyridine-co-styrene) deposited on nitrogen-doped graphene.
Mikrochimica acta (2019-03-25)
Mohammad Saremi, Amir Amini, Hamid Heydari
ABSTRACT
An ultrasensitive electrochemiluminescence (ECL) disposable aptamer sensor (aptasensor) is presented for detection of myocardial infarction biomarker by quantification of troponin I in blood serum. A screen-printed electrode was modified with (a) aptamer-modified gold nanoparticles, (b) cyclometallated iridium(III)-poly-4-vinylpyridine nanoparticles, and (c) nitrogen-doped graphene in order to increase the loading capacity and conductivity of the aptasensor. If the aptasensor is exposed to troponin I, it will bind to the aptamer and desorb the aptamer from gold nanoparticles and the surface of the electrode. This generates an enhancement in ECL emission depending on troponin I concentration. ECL emission is strongly improved by aggregation-induced phenomenon, which is caused by inhibition of the water and oxygen quenching effect on the iridium complex ECL in aqueous media. Under optimum conditions, the aptasensor has a wide dynamic range that extends from 0.1 pM to 10 nM, with a 20 fM detection limit (S/N = 3) and a relative standard deviation of 3.1%. The ECL aptasensor was successfully applied to 20 individual human serum for the detection of troponin I biomarker. Graphical abstract Schematic presentation of electrochemiluminescence aptamer assay fabrication for detection of Troponin I. Carbon screen printed electrode (CSPE) was modified with nitrogen doped graphene (NG), gold nanoparticles (AuNPs), cyclometallated iridium(III)-polyvinylpyridine polymer nanoparticles, ionic liquid and bovine serum albumin.