MilliporeSigma
  • On-off-on relay fluorescence recognition of ferric and fluoride ions based on indicator displacement in living cells.
On-off-on relay fluorescence recognition of ferric and fluoride ions based on indicator displacement in living cells.
Analytica chimica acta (2019-04-28)
Muthusamy Selvaraj, Kanagaraj Rajalakshmi, Yun-Sik Nam, Yeonhee Lee, Jong-Won Song, Ho-Jin Lee, Kang-Bong Lee
ABSTRACT
A new boronic acid derivative functionalized with a 4-(3-(4-(4,5-diphenyl-1H-imidazole-2-yl)phenyl)-1,2,4-oxadiazol-5-yl)phenyl (IOP) moiety was synthesized for use as a sequential "on-off-on"-type relay fluorescence probe for Fe3+ ions and F- ions with high selectivity and sensitivity under physiological conditions. The introduction of Fe3+ to IOP boronic acid (IOPBA) formed an Fe3+IOPBA complex, which led to quenching of the blue fluorescence intensity at 458 nm. The lowest-energy conformation of IOPBA was theoretically predicted to adopt an extended structure, and the Fe3+ ion in the Fe3+IOPBA complex was coordinated to two phenyl groups to form a π-complex. Upon addition of F- to the Fe3+IOPBA complex, the original fluorescence was recovered due to formation of [FeF6]3‒, resulting in "on-off-on"-type sensor behavior. IOPBA showed high selectivity towards Fe3+ among other cations. Moreover, the Fe3+IOPBA complex showed specific selectivity towards F-, with other cations and anions not interfering with detection. Both sensing processes showed 1:1 stoichiometry with binding constants of 6.87 × 106 and 4.49 × 106 mol-1 L for Fe3+ with IOPBA and F- with Fe3+IOPBA, respectively. The limits of detection for Fe3+ and F- were 10 and 1 nM, respectively. The proposed method was successfully applied in real water samples. Furthermore, the probe had low cytotoxicity and was successfully used as a bioimaging reagent to detect intracellular Fe3+ and F- in living HeLa cells.