MilliporeSigma
  • Estrogen-Mediated MicroRNA-101-3p Expression Represses Hyaluronan Synthase 2 in Synovial Fibroblasts From Idiopathic Condylar Resorption Patients.
Estrogen-Mediated MicroRNA-101-3p Expression Represses Hyaluronan Synthase 2 in Synovial Fibroblasts From Idiopathic Condylar Resorption Patients.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2019-03-25)
Chi Feng, Ping Ji, Ping Luo, Jie Xu
ABSTRACT
Idiopathic condylar resorption (ICR) is an aggressive form of osteoarthritis that is frequently observed in adolescent female patients. We hypothesized that an estrogen-mediated pathway may contribute to ICR development. An enzyme-linked immunosorbent assay was used to detect the levels of estradiol (E2) and hyaluronan in synovial fluid. Immunohistochemistry, real-time polymerase chain reaction, and Western blotting were used to detect the expression of microRNAs (miRNAs) and related genes after transfection of miRNA-101-3p mimics, inhibitor, or short interfering RNA into synovial fibroblasts. Dual-luciferase activity was determined to identify the direct effect of miRNA-101-3p on hyaluronan synthase 2 (HAS2). Linear regression analysis, the nonparametric Mann-Whitney U test, the Student t test, and 1-way analysis of variance were carried out to analyze the results of each group. The relationship between hyaluronan and E2 was negatively correlated in synovial fluid (Pearson r = -0.3179, P = .0230). Among the screened miRNAs, miRNA-101-3p was the most overexpressed in ICR. E2 mostly upregulated the expression of miRNA-101-3p at a dose of 10 nmol/L 12 hours after transfection in synovial fibroblasts of patients with ICR. However, E2 induction of miRNA-101-3p expression was significantly repressed by estrogen receptor α interference (P = 0.0286). The dual-luciferase assay showed that miRNA-101-3p regulated the expression of HAS2 by directly targeting its 3' untranslated region. We speculate that E2 regulates HAS2 expression by targeting miRNA-101-3p in synovial fibroblasts of patients with ICR. Thus, the E2-miRNA-101-3p-HAS2 pathway might play an important role in the pathogenesis of ICR.