MilliporeSigma
  • Ultra-sensitive, stable isotope assisted quantification of multiple urinary mycotoxin exposure biomarkers.
Ultra-sensitive, stable isotope assisted quantification of multiple urinary mycotoxin exposure biomarkers.
Analytica chimica acta (2018-04-08)
Bojan Šarkanj, Chibundu N Ezekiel, Paul C Turner, Wilfred A Abia, Michael Rychlik, Rudolf Krska, Michael Sulyok, Benedikt Warth
ABSTRACT
There is a critical need to better understand the patterns, levels and combinatory effects of exposures we are facing through our diet and environment. Mycotoxin mixtures are of particular concern due to chronic low dose exposures caused by naturally contaminated food. To facilitate new insights into their role in chronic disease, mycotoxins and their metabolites are quantified in bio-fluids as biomarkers of exposure. Here, we describe a highly sensitive urinary assay based on ultra-high performance liquid chromatography - tandem mass spectrometer (UHPLC-MS/MS) and 13C-labelled or deuterated internal standards covering the most relevant regulated and emerging mycotoxins. Utilizing enzymatic pre-treatment, solid phase extraction and UHPLC separation, the sensitivity of the method was significantly higher (10-160x lower LODs) than in a previously described method used for comparison purpose, and stable isotopes provided compensation for challenging matrix effects. This method was in-house validated and applied to re-assess mycotoxin exposure in urine samples obtained from Nigerian children, adolescent and adults, naturally exposed through their regular diet. Owing to the methods high sensitivity, biomarkers were detected in all samples. The mycoestrogen zearalenone was the most frequently detected contaminant (82%) but also ochratoxin A (76%), aflatoxin M1 (73%) and fumonisin B1 (71%) were quantified in a large share of urines. Overall, 57% of 120 urines were contaminated with both, aflatoxin M1 and fumonisin B1, and other co-exposures were frequent. These results clearly demonstrate the advanced performance of the method to assess lowest background exposures (pg mL-1 range) using a single, highly robust assay that will allow for the systematic investigation of low dose effects on human health.