Merck
  • Silencing the OCT4-PG1 pseudogene reduces OCT-4 protein levels and changes characteristics of the multidrug resistance phenotype in chronic myeloid leukemia.
Silencing the OCT4-PG1 pseudogene reduces OCT-4 protein levels and changes characteristics of the multidrug resistance phenotype in chronic myeloid leukemia.
Molecular biology reports (2019-02-06)
Aline Portantiolo Lettnin, Eduardo Felipe Wagner, Michele Carrett-Dias, Karina Dos Santos Machado, Adriano Werhli, Andrés Delgado Cañedo, Gilma Santos Trindade, Ana Paula de Souza Votto
ABSTRACT
Cancer stem cells show epigenetic plasticity and intrinsic resistance to anti-cancer therapy, rendering capable of initiating cancer relapse and progression. Transcription factor OCT-4 regulates various pathways in stem cells, but its expression can be regulated by pseudogenes. This work evaluated how OCT4-PG1 pseudogene can affect OCT-4 expression and mechanisms related to the multidrug resistance (MDR) phenotype in FEPS cells. Considering that OCT-4 protein is a transcription factor that regulates expression of ABC transporters, level of gene expression, activity of ABC proteins and cell sensitivity to chemotherapy were evaluated after OCT4-PG1 silencing. Besides we set up a STRING network. Results showed that after OCT4-PG1 silencing, cells expressed OCT-4 gene and protein to a lesser extent than mock cells. The gene and protein expression of ABCB1, as well as its activity were reduced. On the other hand, ALOX5 and ABCC1 genes was increased even as the activity of this transporter. Moreover, the silencing cells become sensitive to two chemotherapics tested. The network structure demonstrated that OCT4-PG1 protein interacts directly with OCT-4, SOX2, and NANOG and indirectly with ABC transporters. We conclude that OCT4-PG1 pseudogene plays a key role in the regulation OCT-4 transcription factor, which alters MDR phenotype in the FEPS cell line.