MilliporeSigma
  • Qualitative and quantitative determination of Atractylodes rhizome using ultra-performance liquid chromatography coupled with linear ion trap-Orbitrap mass spectrometry with data-dependent processing.
Qualitative and quantitative determination of Atractylodes rhizome using ultra-performance liquid chromatography coupled with linear ion trap-Orbitrap mass spectrometry with data-dependent processing.
Biomedical chromatography : BMC (2018-11-24)
Yihan Zhang, Cui Bo, Yanhao Fan, Rui An, Long Chen, Yifan Zhang, Yiqun Jia, Xinhong Wang
ABSTRACT
A quick and effective workflow based on ultra-performance liquid chromatography coupled with electron spray ionization and LTQ-Orbitrap mass spectrometry (UPLC-LTQ-Orbitrap MS) was established for compositional analysis and screening of the characteristic compounds of three species of Atractylodes rhizome for quality evaluation. This technique was employed to determine the seven main components in Atractylodes rhizome samples. Ultimately, 78 constituents were identified; of these, seven characteristic compounds were selected for species discrimination, comprising atractylodin (63), atractylenolide I (43), atractylenolide II (49), atractylenolide III (53), atractylon (69), methyl-atractylenolide II (54) and (4E,6E,12E)-tetradecadecatriene-8,10-diyne-1,3-diacetate (59). The seven main compounds, including six characteristic compounds, were simultaneously determined in 29 batches of Atractylodes rhizome samples. Thus, the method validation showed acceptable results. Quantitative analysis showed significantly different contents of the seven main components among the three species of Atractylodes rhizome, which indicates possible distinctions in the pharmacological effects. This established method can simultaneously provide qualitative and quantitative results for compositional characterization of Atractylodes rhizomes and for quality control.