MilliporeSigma
  • Synthesis and antimicrobial activity of a phenanthroline-isoniazid hybrid ligand and its Ag+ and Mn2+ complexes.
Synthesis and antimicrobial activity of a phenanthroline-isoniazid hybrid ligand and its Ag+ and Mn2+ complexes.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (2019-06-24)
Muhib Ahmed, Denise Rooney, Malachy McCann, Michael Devereux, Brendan Twamley, Anna Clara Milesi Galdino, Leandro Stefano Sangenito, Lucieri Olegario Pereira Souza, Maria Cristina Lourenço, Karen Gomes, André Luis Souza Dos Santos
ABSTRACT
Hydrazide ligand, (Z)-N'-(6-oxo-1,10-phenanthrolin-5(6H)-ylidene)isonicotinohydrazide, 1 forms from a 1:1 Schiff base condensation reaction between isoniazid (INH) and 1,10-phenanthroline-5,6-dione (phendione). Ag+ and Mn2+ complexes with 1:2 metal:ligand stoichiometry are prepared: [Ag(1)2]NO3, [Ag(1)2]BF4 and [Mn(1)2](NO3)2. Polymeric {[Ag(1)(NO3)]}n has 1:1 stoichiometry and forms upon infusion of CH2Cl2 into a DMSO solution of [Ag(1)2]NO3. {[Ag(1)(NO3)]}n was structurally characterized using X-ray crystallography. Metal-free 1 and its 1:2 complexes exhibit very good, broad-spectrum antimicrobial activity and are not excessively toxic to mammalian cells (A549 lineage).