MilliporeSigma
  • A composite of platelet-like orientated BiVO4 fused with MIL-125(Ti): Synthesis and characterization.
A composite of platelet-like orientated BiVO4 fused with MIL-125(Ti): Synthesis and characterization.
Scientific reports (2019-07-13)
Philani Vusumuzi Hlophe, Lwazi Charles Mahlalela, Langelihle Nsikayezwe Dlamini
ABSTRACT
The development of heterojunctions is the current focus of the scientific community as these materials are visible light active and the staggered positioning of their band edges combats electron-hole recombination which is the downside of most photocatalysts. In this work, a two- step hydrothermal synthesis protocol was utilized to fabricate a novel observable-light active material, composed of platelet-like BiVO4 and a titanium-based metal organic framework (MOF) called MIL-125(Ti). The tuning of specific morphologies, such as platelet-like in BiVO4, provides the exposure of most reactive facets which are more reactive towards photooxidation of organics in water, thus increasing their efficiency. The as-synthesized heterojunction was characterized by Transmission electron microscopy (TEM), scanning transmission microscopy (SEM), X-Ray diffraction (XRD), Raman spectroscopy, ultraviolet-visible diffuse reflectance spectra (UV-Vis DRS), X-Ray photoelectron spectroscopy (XPS) and photoluminescence (PL) spectra. The formation of the heterojunction lead to a positive shift of the 3-2 Bi:Ti valence band (VB) (1.78 eV) when compared to 1.27 eV VB position of BiVO4. The PL and photoelectrochemical measurements revealed that the heterojunction photocatalyst designated 3-2 Bi-Ti demonstrated inhibited recombination rate (platelet-like BiVO4 > 3-2 Bi:Ti (PM) > MIL-125 > 1-1 Bi:Ti > 2-3 Bi:Ti > 3-2 Bi:Ti) and highly efficient interfacial charge shuttle between platelet-like BiVO4 and MIL-125(Ti) through the formed n-n junction.