MilliporeSigma
  • Sensitivity of glutathione S-transferases to high doses of acrylamide in albino wistar rats: Affinity purification, biochemical characterization and expression analysis.
Sensitivity of glutathione S-transferases to high doses of acrylamide in albino wistar rats: Affinity purification, biochemical characterization and expression analysis.
Ecotoxicology and environmental safety (2019-07-14)
Sreenivasulu Dasari, Sailaja Gonuguntla, Prabhakar Yellanurkonda, Prabhusaran Nagarajan, Balaji Meriga
ABSTRACT
The main objectives of this study were to purify the glutathione S-transfereses (GSTs) and assess the effect of high doses of acrylamide (ACR) on male albino Wistar rat liver, kidney, testis and bran GST activities, and expression analysis of GST. ACR (50 mg/300 ml) was ingested for 40 days (20 doses) in drinking water on alternative days, on 40 day post ingestion the control and treated tissues were collected for GST purification by affinity column and biochemical characterization of GSTs by substrate specificities, and GST expression by immuno dot blots. In the analysis of the purified GSTs, we observed that liver GSTs were resolved in to three bands known as Yc, Yb and Ya; kidney GSTs were resolved in to two bands known as Yc and Ya; testis and brain GSTs were resolved as four bands known as Yc, Yb, Yβ and Yδ on 12.5% sodium dodecyl sulfate polyacrylamide gel (SDS PAGE). In the analysis of biochemical characterization, we observed a significant decrease (p < 0.05) in the specific activities of liver GST isoforms with the substrates 1-chloro 2,4-dinitrobenzene (CDNB), bromosulfophthalein (BSP), p-nitrophenyl acetate (pNPA), p-nitrobenzyl chloride (pNBC) and cumene hydroperoxide (CHP), but showed no activity with ethacrynic acid (ECA) and significant decrease (p < 0.05) in the specific activities of kidney GST isoforms with the substrates CDNB, pNPA, pNBC and CHP, but showed no activity with BSP and ECA, and a significant decrease (p < 0.05) in the specific activities of testis and brain GST isoforms with the substrates CDNB, BSP, pNPA, pNBC, ECA and CHP. In the analysis of immuno dot blots, we observed a decreased expression of liver, kidney, testis and brain GSTs. Through the affinity purification and biochemical characterization, we observed a tissue specific distribution of GSTs that is liver GSTs possess Yc, Yb and Ya sub units known as alpha (α) and mu (μ) class GSTs; kidney GSTs possess Yc and Ya sub units known as (α) alpha class GST; testis and brain GSTs possess Yc, Yb, Yβ and Yδ sub units known as alpha (α), mu (μ) and pi (π) class GSTs. Purification studies, biochemical characterization and immuno dot blot analysis were revealed the GSTs were sensitive to high doses of ACR and the high level exposure to ACR cause the damage of detoxification function of GST due to decreased expression and hence lead to cellular dysfunction of vital organs.