MilliporeSigma
  • Retrodifferentiation of Human Tumor Hepatocytes to Stem Cells Leads to Metabolic Reprogramming and Chemoresistance.
Retrodifferentiation of Human Tumor Hepatocytes to Stem Cells Leads to Metabolic Reprogramming and Chemoresistance.
Cancer research (2019-03-07)
Karim Fekir, Hélène Dubois-Pot-Schneider, Romain Désert, Yoann Daniel, Denise Glaise, Claudine Rauch, Fabrice Morel, Bernard Fromenty, Orlando Musso, Florian Cabillic, Anne Corlu
ABSTRACT
Human hepatocellular carcinoma (HCC) heterogeneity promotes recurrence and therapeutic resistance. We recently demonstrated that inflammation favors hepatocyte retrodifferentiation into progenitor cells. Here, we identify the molecular effectors that induce metabolic reprogramming, chemoresistance, and invasiveness of retrodifferentiated HCC stem cells. Spheroid cultures of human HepaRG progenitors (HepaRG-Spheres), HBG-BC2, HepG2, and HuH7 cells and isolation of side population (SP) from HepaRG cells (HepaRG-SP) were analyzed by transcriptomics, signaling pathway analysis, and evaluation of chemotherapies. Gene expression profiling of HepaRG-SP and HepaRG-Spheres revealed enriched signatures related to cancer stem cells, metastasis, and recurrence and showed that HepaRG progenitors could retrodifferentiate into an immature state. The transcriptome from these stem cells matched that of proliferative bad outcome HCCs in a cohort of 457 patients. These HCC stem cells expressed high levels of cytokines triggering retrodifferentiation and displayed high migration and invasion potential. They also showed changes in mitochondrial activity with reduced membrane potential, low ATP production, and high lactate production. These changes were, in part, related to angiopoietin-like 4 (ANGPTL4)-induced upregulation of pyruvate dehydrogenase kinase 4 (PDK4), an inhibitor of mitochondrial pyruvate dehydrogenase. Upregulation of ANGPTL4 and PDK4 paralleled that of stem cells markers in human HCC specimens. Moreover, the PDK4 inhibitor dichloroacetate reversed chemoresistance to sorafenib or cisplatin in HCC stem cells derived from four HCC cell lines. In conclusion, retrodifferentiated cancer cells develop enhanced invasion and therapeutic resistance through ANGPTL4 and PDK4. Therefore, restoration of mitochondrial activity in combination with chemotherapy represents an attractive therapeutic approach in HCC. SIGNIFICANCE: Restoring mitochondrial function in human hepatocellular carcinomas overcomes cancer resistance.
Sigma-Aldrich
Anti-LGR5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HAPLN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1