MilliporeSigma
  • Extracellular Hsp90α and clusterin synergistically promote breast cancer epithelial-to-mesenchymal transition and metastasis via LRP1.
Extracellular Hsp90α and clusterin synergistically promote breast cancer epithelial-to-mesenchymal transition and metastasis via LRP1.
Journal of cell science (2019-07-06)
Yang Tian, Chunying Wang, Shuohua Chen, Jie Liu, Yan Fu, Yongzhang Luo
ABSTRACT
Extracellular heat shock protein 90 alpha (eHsp90α, also known as HSP90AA1) has been widely reported to promote tumor cell motility and tumor metastasis in various types of cancer. Several extracellular proteins and membrane receptors have been identified as interacting proteins of eHsp90α and mediate its pro-metastasis function. However, the regulatory mechanism of eHsp90α activity remains largely unknown. Here, we report that clusterin, a protein newly demonstrated to interact with eHsp90α, modulates eHsp90α signaling. We found that clusterin potentiated the effects of eHsp90α on activation of the AKT, ERK and NF-κB protein families, epithelial-to-mesenchymal transition (EMT) and migration in breast cancer cells. Furthermore, in vivo investigations demonstrated similar synergistic effects of eHsp90α and clusterin on tumor metastasis. Notably, the effects of eHsp90α and clusterin were mediated by low-density lipoprotein receptor-related protein 1 (LRP1). Proximity ligation assay and co-immunoprecipitation experiments demonstrated that clusterin participated in eHsp90α-LRP1 complex formation, which enhanced the binding affinity of eHsp90α to LRP1. Collectively, our data establish a role of clusterin as a newly discovered modulator of eHsp90α, and unravel detailed molecular mechanisms underlying the synergistic metastasis-promoting effects of clusterin and eHsp90α.
Sigma-Aldrich
Anti-LRP1 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution