MilliporeSigma
  • Increased frequency of psychosis after second-generation antiepileptic drug administration in adults with focal epilepsy.
Increased frequency of psychosis after second-generation antiepileptic drug administration in adults with focal epilepsy.
Epilepsy & behavior : E&B (2019-06-30)
Naoto Adachi, Peter Fenwick, Nozomi Akanuma, Koichiro Hara, Ryouhei Ishii, Mitsutoshi Okazaki, Masumi Ito, Masanori Sekimoto, Masaaki Kato, Teiichi Onuma
ABSTRACT
Many studies show psychoses after some antiepileptic drug (AED) administrations (post-AED administration psychoses [PAP]). It remains uncertain about psychogenetic potential of each AED and effects of clinical state factors on PAP. We examined the relations between AED-related factors (types, generations, dosages, and concomitant AED) and PAP. The clinical records of patients with focal epilepsy were retrospectively reviewed from eight adult epilepsy clinics, for every six-month period after administration of a new drug (either AED or non-AED) between 1981 and 2015. Characteristics of psychotic episodes, AED-related factors (type, daily dosage, and concomitant AED), and other state-related risk factors to psychosis (age, duration of epilepsy, history of psychosis, and seizure frequency) were examined. Psychogenetic risks of AED-related and state-related factors were analyzed with multifactorial procedures. Of 2067 patients with focal epilepsy, 5018 new drugs (4402 AEDs and 616 non-AEDs) were administered. Within the first six-month period, 89 patients exhibited 105 psychotic episodes (81 interictal and 24 postictal psychoses: 55 first episodes and 50 recurrences). With second-generation AED (SAED) administration, particularly topiramate and lamotrigine, frequency of psychosis was significantly increased. Daily dosage of AED was not significantly associated with psychosis. Psychosis tended to occur with a higher number of concomitant AED. Subsequent analysis with AED-related and general factors showed that SAED administrations and previous psychotic history were the most significant risks for PAP. Post-AED administration psychoses is associated with type of AED (SAED), rather than its dosage. Individual vulnerabilities are also associated with PAP.