MilliporeSigma
  • Synthesis and Characterization of Room Temperature Vulcanized Silicone Rubber Using Methoxyl-Capped MQ Silicone Resin as Self-Reinforced Cross-Linker.
Synthesis and Characterization of Room Temperature Vulcanized Silicone Rubber Using Methoxyl-Capped MQ Silicone Resin as Self-Reinforced Cross-Linker.
Polymers (2019-07-07)
Jianye Ji, Xin Ge, Xiaoyan Pang, Ruoling Liu, Shuyi Wen, Jiaqi Sun, Weijie Liang, Jianfang Ge, Xunjun Chen
ABSTRACT
Methoxyl-capped MQ silicone resin (MMQ) was first synthesized by the hydrosilylation of vinyl-containing MQ silicone resin and trimethoxysilane and then used in condensed room-temperature vulcanized (RTV) silicone rubber as a self-reinforced cross-linker. Results show that modified silicone rubber exhibits good light transmission. Compared with unmodified silicone rubber, the hardness, tensile strength and elongation of MMQ at the break are increased by 26.4 A, 2.68 MPa and 65.1%, respectively. In addition, the characteristic temperature of 10% mass loss is delayed from 353.5 °C to 477.1 °C, the temperature at maximum degradation rate is also delayed from 408.9 °C to 528.4 °C and the residual mass left at 800 °C is increased from 1.2% to 27.7%. These improved properties are assigned to the synergistic effect of the rigid structure of MMQ, the formation of a dense cross-linking structure in polymers and the uniform distribution of MMQ cross-linking agent in RTV silicone rubber.