MilliporeSigma
  • Genome-wide mapping reveals that deoxyuridine is enriched in the human centromeric DNA.
Genome-wide mapping reveals that deoxyuridine is enriched in the human centromeric DNA.
Nature chemical biology (2018-05-23)
Xiaoting Shu, Menghao Liu, Zhike Lu, Chenxu Zhu, Haowei Meng, Sihao Huang, Xiaoxue Zhang, Chengqi Yi
ABSTRACT
Uracil in DNA can be generated by cytosine deamination or dUMP misincorporation; however, its distribution in the human genome is poorly understood. Here we present a selective labeling and pull-down technology for genome-wide uracil profiling and identify thousands of uracil peaks in three different human cell lines. Surprisingly, uracil is highly enriched at the centromere of the human genome. Using mass spectrometry, we demonstrate that human centromeric DNA contains a higher level of uracil. We also directly verify the presence of uracil within two centromeric uracil peaks on chromosomes 6 and 11. Moreover, centromeric uracil is preferentially localized within the binding regions of the centromere-specific histone CENP-A and can be excised by human uracil-DNA glycosylase UNG. Collectively, our approaches allow comprehensive analysis of uracil in the human genome and provide robust tools for mapping and future functional studies of uracil in DNA.