MilliporeSigma
  • Promotion of Tumor Growth by ADAMTS4 in Colorectal Cancer: Focused on Macrophages.
Promotion of Tumor Growth by ADAMTS4 in Colorectal Cancer: Focused on Macrophages.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-04-26)
Jianjun Chen, Yang Luo, Yong Zhou, Shaolan Qin, Yier Qiu, Ran Cui, Minhao Yu, Jun Qin, Ming Zhong
ABSTRACT
ADAMTSs (A disintegrin and metalloprotease domains with thrombospondins motifs) are a family of extracellular proteases that have been related to both oncogenic and tumor-suppressive functions. The aim of the present study was to investigate: 1) the mutation, copy-number alterations, and expression profile of ADAMTSs in colorectal cancer and 2) whether ADAMTSs participate in colorectal cancer (CRC) progression and invasion. The mutation, copy-number alterations, and expression profile of ADAMTSs in CRC were analyzed in the TCGA cohort using cBioportal. ADAMTS4 expression in tumor tissues and cell lines were determined by immunostaining and real-time quantitative PCR. The role of ADAMTS-4 in CRC progression and the underlying mechanisms were studied by using short hairpin RNA-mediated knockdown of ADAMTS4. The effects of ADAMTS4 in cell proliferation and invasion were determined by clone formation assay and transwell migration assay, respectively. Macrophages were depleted by liposomal clodronate in immune-competent BALB/c mice and tumor growth was analyzed. ADAMTS4 was differentially expressed in CRC and predicted a poor prognosis. Elevated ADAMTS4 expression was closely associated with larger tumor size, enhanced TNM stage, and a poor clinical outcome in patients with CRC. ADAMTS4 knockdown had no inhibitory implications on cell proliferation and invasion in vitro, but significantly attenuated tumor growth in vivo. Mechanistically, we revealed that ADAMTS4 was associated macrophages infiltration and polarization in the tumor microenvironment of CRC. Macrophage depletion largely abolished the promotive effect of ADAMTS4 on tumor growth in the immune competent BALB/c mice. ADAMTS4 seemed to be a promising prognostic indicator in CRC. The novel link between ADAMTS4 and macrophages mirrors the potential regulatory roles of ADAMTSs in the inflammatory microenvironment of cancers.