MilliporeSigma
  • Imidazole-pyrazole hybrids: Synthesis, characterization and in-vitro bioevaluation against α-glucosidase enzyme with molecular docking studies.
Imidazole-pyrazole hybrids: Synthesis, characterization and in-vitro bioevaluation against α-glucosidase enzyme with molecular docking studies.
Bioorganic chemistry (2018-11-06)
Faryal Chaudhry, Sadia Naureen, Muhammad Ashraf, Mariya Al-Rashida, Bakhat Jahan, Munawar Ali Munawar, Misbahul Ain Khan
ABSTRACT
Herein, substituted imidazole-pyrazole hybrids (2a-2n) were prepared via a multi component reaction employing pyrazole-4-carbaldehydes (1a-1d), ammonium acetate, benzil and arylamines as reactants. All the new compounds were characterized through their spectral and elemental analyses. Further these compounds were tested against α-glucosidase enzyme. The compounds 2k, 2l and 2n possessed good inhibition potencies, however, compounds 2f (IC50 value: 25.19 ± 0.004 μM) and 2m (IC50 value: 33.62 ± 0.03 μM) were the most effective compounds of the series. Furthermore, molecular docking helped to understand the binding interactions of 2f and 2m with the understudy yeast's α-glucosidase enzyme.