MilliporeSigma
  • Ultrasound-mediated destruction of vascular endothelial growth factor (VEGF) targeted and paclitaxel loaded microbubbles for inhibition of human breast cancer cell MCF-7 proliferation.
Ultrasound-mediated destruction of vascular endothelial growth factor (VEGF) targeted and paclitaxel loaded microbubbles for inhibition of human breast cancer cell MCF-7 proliferation.
Molecular and cellular probes (2019-06-23)
Jilian Su, Junmei Wang, Jiamin Luo, Haili Li
ABSTRACT
Vascular endothelial growth factor (VEGF) can promote cell division, proliferation and migration. In this study, we aimed to investigate roles of ultrasound-mediated destruction of VEGF-targeted and paclitaxel (PTX)-loaded lipid microbubbles (VTPLLM + US) in human breast cancer cells. The activity of MCF-7 cells was determined by cell counting Kit-8. Flow cytometry was performed to detect the cells apoptosis and cell cycle. The expression of cell cycle-associated proteins, matrix metalloprotein-9 (MMP-9), VEGF and apoptosis-associated proteins were detected by qRT-PCR and Western blot. The obtained data suggested that VTPLLM + US promoted the G1 phase cell cycle arrest and suppressed the viability of MCF-7 cells. We also found that VTPLLM + US accelerated cells apoptosis. Cell cycle-associated proteins and VEGF expression were modulated by VTPLLM + US. Moreover, VTPLLM + US was found to regulate the expression levels of apoptosis-associated proteins in MCF-7 cells. Our findings suggested that VTPLLM + US suppressed the proliferation and accelerated the apoptosis of MCF-7 cells through regulating VEGF expression. The potential effects of VTPLLM + US on apoptosis of MCF-7 cells suggest that applying VTPLLM + US might be an effective strategy in breast cancer therapies.