MilliporeSigma
  • Wound healing activity and phytochemical screening of purified fractions of Sempervivum tectorum L. leaves on HCT 116.
Wound healing activity and phytochemical screening of purified fractions of Sempervivum tectorum L. leaves on HCT 116.
Phytochemical analysis : PCA (2019-06-07)
Fabio Cattaneo, Simona De Marino, Melania Parisi, Carmen Festa, Martina Castaldo, Claudia Finamore, Francesca Duraturo, Cristiana Zollo, Rosario Ammendola, Franco Zollo, Maria Iorizzi
ABSTRACT
Sempervivum tectorum L. (Crassulaceae), is a succulent perennial plant widespread in Mediterranean countries and commonly used in traditional medicine for ear inflammation, ulcers and skin rashes as a refrigerant and astringent. To demonstrate the therapeutic effects of the plant, various fractions were purified and characterised. The potential wound healing activity, proliferation rate and intracellular signalling cascades were investigated by using human epithelial colorectal carcinoma (HCT 116) cells. An extraction method without organic solvents was applied for the first time. The purification was carried out by droplet counter current chromatography (DCCC) coupled with high-performance liquid chromatography (HPLC) and electrospray ionisation mass spectrometry (ESI-MS) data. By nuclear magnetic resonance (NMR) [1 H, 13 C and two-dimensional (2D) experiments] pure components were identified. Wound healing and cell proliferation assays were utilised to determine the role of the isolated S. tectorum (SVT) fraction on cellular migration and proliferation. The signalling pathways elicited from the SVT fractions, were analysed by Western blot analysis. In this study two rare natural components were identified, namely monosaccharide sedoheptulose and polyalcohol 2-C-methyl-D-erythritol, along with known organic acids and flavonoids. The fractions with high level of sedoheptulose enhance the proliferation and the cellular migration of epithelial HCT 116 cells. The intracellular signalling cascades elicited from the purified fractions induce the c-Src-mediated transactivation of EGFR and the activation of the STAT3 pathway which, in turn, are crucially involved in the cellular proliferation and migration. Our study demonstrates the efficacy of purified fractions of S. tectorum L. in enhancing cellular proliferation and migration, suggesting their potential role as topical therapeutic treatments for wound healing.