MilliporeSigma
  • CRISPR-Cas9 Circular Permutants as Programmable Scaffolds for Genome Modification.
CRISPR-Cas9 Circular Permutants as Programmable Scaffolds for Genome Modification.
Cell (2019-01-12)
Benjamin L Oakes, Christof Fellmann, Harneet Rishi, Kian L Taylor, Shawn M Ren, Dana C Nadler, Rayka Yokoo, Adam P Arkin, Jennifer A Doudna, David F Savage
ABSTRACT
The ability to engineer natural proteins is pivotal to a future, pragmatic biology. CRISPR proteins have revolutionized genome modification, yet the CRISPR-Cas9 scaffold is not ideal for fusions or activation by cellular triggers. Here, we show that a topological rearrangement of Cas9 using circular permutation provides an advanced platform for RNA-guided genome modification and protection. Through systematic interrogation, we find that protein termini can be positioned adjacent to bound DNA, offering a straightforward mechanism for strategically fusing functional domains. Additionally, circular permutation enabled protease-sensing Cas9s (ProCas9s), a unique class of single-molecule effectors possessing programmable inputs and outputs. ProCas9s can sense a wide range of proteases, and we demonstrate that ProCas9 can orchestrate a cellular response to pathogen-associated protease activity. Together, these results provide a toolkit of safer and more efficient genome-modifying enzymes and molecular recorders for the advancement of precision genome engineering in research, agriculture, and biomedicine.
Sigma-Aldrich
Anti-CRISPR/Cas9 (C-terminal) antibody, Mouse monoclonal, clone 10C11-A12, purified from hybridoma cell culture
Pricing and availability is not currently available.