MilliporeSigma
  • Suppression of proliferation and activation of cell death by sodium selenite involves mitochondria and lysosomes in chemoresistant bladder cancer cells.
Suppression of proliferation and activation of cell death by sodium selenite involves mitochondria and lysosomes in chemoresistant bladder cancer cells.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS) (2019-02-09)
K Soukupová, E Rudolf
ABSTRACT
The specific effects of sodium selenite (selenite) on a chemoresistant human bladder cancer cell line RT-112/D21 were investigated during 72 h. Selenite at low concentration of 2.5 μmol (otherwise tolerated in normal urothelial cells UROtsa) suppressed growth and proliferation of the tested cancer cells via induced oxidative stress. Selenite further altered mitochondrial functions (i.e. decreased mitochondrial membrane potential, increased production of superoxide and reduced ATP synthesis), disrupted lysosomal membranes and activated autophagy. These changes in selenite-exposed cells ultimately resulted in their demise via necrosis and other cell death modality displaying heterotypic apoptotic and autophagic features.