MilliporeSigma
  • Adaptation of Bacillus species to dairy associated environment facilitates their biofilm forming ability.
Adaptation of Bacillus species to dairy associated environment facilitates their biofilm forming ability.
Food microbiology (2019-04-28)
Ievgeniia Ostrov, Noa Sela, Eduard Belausov, Doron Steinberg, Moshe Shemesh
ABSTRACT
Biofilm-forming Bacillus species are often involved in contamination of dairy products and therefore present a major microbiological challenge in the field of food quality and safety. In this study, we sequenced and analyzed the genomes of milk- and non-milk-derived Bacillus strains, and evaluated their biofilm-formation potential in milk. Unlike non-dairy Bacillus isolates, the dairy-associated Bacillus strains were characterized by formation of robust submerged and air-liquid interface biofilm (pellicle) during growth in milk. Moreover, genome comparison analysis revealed notable differences in putative biofilm-associated determinants between the dairy and non-dairy Bacillus isolates, which correlated with biofilm phenotype. These results suggest that biofilm formation by Bacillus species might represent a presumable adaptation strategy to the dairy environment.