MilliporeSigma
  • Wafer-scale single-crystal hexagonal boron nitride film via self-collimated grain formation.
Wafer-scale single-crystal hexagonal boron nitride film via self-collimated grain formation.
Science (New York, N.Y.) (2018-11-18)
Joo Song Lee, Soo Ho Choi, Seok Joon Yun, Yong In Kim, Stephen Boandoh, Ji-Hoon Park, Bong Gyu Shin, Hayoung Ko, Seung Hee Lee, Young-Min Kim, Young Hee Lee, Ki Kang Kim, Soo Min Kim
ABSTRACT
Although polycrystalline hexagonal boron nitride (PC-hBN) has been realized, defects and grain boundaries still cause charge scatterings and trap sites, impeding high-performance electronics. Here, we report a method of synthesizing wafer-scale single-crystalline hBN (SC-hBN) monolayer films by chemical vapor deposition. The limited solubility of boron (B) and nitrogen (N) atoms in liquid gold promotes high diffusion of adatoms on the surface of liquid at high temperature to provoke the circular hBN grains. These further evolve into closely packed unimodal grains by means of self-collimation of B and N edges inherited by electrostatic interaction between grains, eventually forming an SC-hBN film on a wafer scale. This SC-hBN film also allows for the synthesis of wafer-scale graphene/hBN heterostructure and single-crystalline tungsten disulfide.