MilliporeSigma
  • Gas chromatography-isotope dilution mass spectrometry method validation for target pesticides in soybeans.
Gas chromatography-isotope dilution mass spectrometry method validation for target pesticides in soybeans.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2018-12-14)
J F Huertas-Pérez, B Sejerøe-Olsen, T Gokcen, F Sandor, H Schimmel, M Dabrio
ABSTRACT
The production of certified reference materials requires the application of highly accurate methods for characterisation. A gas chromatography-isotope dilution mass spectrometry method, setting ambitious performance criteria, was developed for eight selected pesticides in soybeans. Pressurised liquid extraction was followed by automated gel-permeation chromatography and solid-phase extraction clean-up. Pesticides identification respected a Commission Decision and guidelines of the Directorate General for Health and Food Safety (DG SANTE). Reliable quantification involved stable isotopically labelled analogues as internal standards. Validation, according to ISO/IEC 17,025 and DG SANTE guidelines, assessed linearity, LOD/LOQ, trueness, selectivity, precision, stability and robustness. Mean recoveries ranges (83-109%, relative standard deviations < 3%), repeatability (2.2-4.8%), day-to-day variation (0.6-4.2%) and combined uncertainty (1.2-4.2%) were fit for purpose. The method is highly accurate and suitable for certification of the selected pesticides in soybean matrix reference material. Chemical compounds studied in this article: Diazinon (PubChem CID: 3017); malathion (PubChem CID: 4004); chlorpyrifos (PubChem CID: 2730); captan (PubChem CID: 8606); endosulfan (PubChem CID: 3224); tebuconazole (PubChem CID: 86,102); iprodione (PubChem CID: 37,517); cypermethrin (PubChem CID: 2912).