MilliporeSigma
  • Rh2(II)-catalyzed intramolecular aliphatic C-H bond amination reactions using aryl azides as the N-atom source.
Rh2(II)-catalyzed intramolecular aliphatic C-H bond amination reactions using aryl azides as the N-atom source.
Journal of the American Chemical Society (2012-04-24)
Quyen Nguyen, Ke Sun, Tom G Driver
ABSTRACT
Rhodium(II) dicarboxylate complexes were discovered to catalyze the intramolecular amination of unactivated primary, secondary, or tertiary aliphatic C-H bonds using aryl azides as the N-atom precursor. While a strong electron-withdrawing group on the nitrogen atom is typically required to achieve this reaction, we found that both electron-rich and electron-poor aryl azides are efficient sources for the metal nitrene reactive intermediate.
Sigma-Aldrich
Phenylboronic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Phenylboronic acid, 95%
Sigma-Aldrich
trans-2-Phenylvinylboronic acid, 97%