MilliporeSigma
  • miRNA-31-5p Mediates the Proliferation and Apoptosis of Human Spermatogonial Stem Cells via Targeting JAZF1 and Cyclin A2.
miRNA-31-5p Mediates the Proliferation and Apoptosis of Human Spermatogonial Stem Cells via Targeting JAZF1 and Cyclin A2.
Molecular therapy. Nucleic acids (2018-12-26)
Hongyong Fu, Fan Zhou, Qingqing Yuan, Wenhui Zhang, Qianqian Qiu, Xing Yu, Zuping He
ABSTRACT
Several lines of evidence highlight the important application of human spermatogonial stem cells (SSCs) in translational medicine. The fate decisions of SSCs are mainly mediated by genetic and epigenetic factors. We have recently demonstrated that PAK1 regulates the proliferation, DNA synthesis, and early apoptosis of human SSCs through the PDK1/KDR/ZNF367 and ERK1/2 and AKT pathway. However, the underlying epigenetic mechanism of PAK1 in human SSCs remains unknown. In this study, we found that the level of miRNA-31-5p was elevated by PAK1 knockdown. CCK-8, PCNA, and 5-ethynyl-2'-deoxyuridine (EDU) assays revealed that miRNA-31-5p mimics inhibited cell proliferation and DNA synthesis of human SSCs. Annexin V/propidium iodide (PI) staining and flow cytometry showed that miRNA-31-5p increased the early and late apoptosis of human SSCs. Furthermore, JAZF1 was predicted and verified as a target of miRNA-31-5p, and the three-dimensional (3D) structure model of JAZF1 protein was illustrated. JAZF1 silencing led to a reduction of cell proliferation and DNA synthesis as well as an enhancement of the early and late apoptosis of human SSCs. Finally, miRNA-31-5p mimics decreased the level of cyclin A2 rather than cyclin D1 or cyclin E1, and JAZF1 knockdown led to the reduction of cyclin A2 in human SSCs. Collectively, miRNA-31-5p regulates the proliferation, DNA synthesis, and apoptosis of human SSCs by the PAK1-JAZF1-cyclin A2 pathway. This study thus offers a novel insight into the molecular mechanisms underlying the fate determinations of human SSCs and might provide novel targets for molecular therapy of male infertility.