MilliporeSigma
  • Bifunctional MIL-53(Fe) with pyrophosphate-mediated peroxidase-like activity and oxidation-stimulated fluorescence switching for alkaline phosphatase detection.
Bifunctional MIL-53(Fe) with pyrophosphate-mediated peroxidase-like activity and oxidation-stimulated fluorescence switching for alkaline phosphatase detection.
Journal of materials chemistry. B (2019-08-08)
Kun Ye, Linjie Wang, Hongwei Song, Xin Li, Xiangheng Niu
ABSTRACT
Alkaline phosphatase (ALP) is extensively used as a clinical biomarker because of its close relevance with a variety of diseases. Thus, exploring reliable and practical methods for ALP analysis is of great significance. In the present work, we explored MIL-53(Fe) as a bifunctional platform with pyrophosphate (PPi)-mediated peroxidase-like activity and oxidation-stimulated fluorescence switching for ALP sensing. The proposed MIL-53(Fe) could exhibit favorable peroxidase-mimicking activity to catalytically decompose H2O2 to hydroxyl radicals, which had strong oxidizing ability to oxidize the terephthalic acid bridging ligand, resulting in the oxidation-stimulated turn-on fluorescence of MIL-53(Fe) itself. Due to the strong coordination interaction between PPi and Fe3+, the former with a relatively large molecular structure was able to inhibit the catalytic activity of MIL-53(Fe) via capping active Fe3+ sites, leading to the suppression of its self-fluorescence response. When ALP was present, it could hydrolyze the PPi inhibitor and restore the dual functions of MIL-53(Fe) to provide fluorescence again. With the above principle, highly sensitive and selective determination of ALP with a linear scope of 2-80 U L-1 and a detection limit down to 0.7 U L-1 was achieved. The MIL-53(Fe) was also demonstrated to be very reliable in measuring the target in human serum, indicating its great promise as an integrated tool for ALP detection in clinical practice.