MilliporeSigma
  • Impairment of rat oesophageal muscle contractility associated with experimental non-erosive oesophageal mucosal damage.
Impairment of rat oesophageal muscle contractility associated with experimental non-erosive oesophageal mucosal damage.
Experimental physiology (2018-12-19)
Kalinne Kelly Lima Gadelha, Francisco José Batista-Lima, Daniel Maia Nogueira de Oliveira, Emanuella Feitosa de Carvalho, Daniel Sifrim, Armênio Aguiar Dos Santos, Pedro Jorge Caldas Magalhães
ABSTRACT
What is the central question of this study? Is the responsiveness of isolated segments of the rat oesophagus to contractile or relaxant stimuli susceptible to acute luminal exposure of the oesophagus to an acid solution that contains pepsin and bile salt? What is the main finding and its importance? The study reveals that luminal acidity is an important factor that disrupts barrier function in the oesophagus to allow the diffusion of noxious agents, such as bile acid, that alter the contractile status of the oesophageal body, even in the absence of inflammation. We investigated whether the experimental simulation of duodenogastro-oesophageal reflux alters the contractile responsiveness of rat oesophageal strips. After 30 min of luminal exposure to a solution at acid pH that contained pepsin and taurodeoxycholic acid, isolated strips of the rat oesophagus and gastro-oesophageal junction were subjected to contractile or relaxing stimuli. Acid challenge decreased the responsiveness of oesophageal strips to contractile stimulation, especially in oesophageal preparations that were mounted following the circular orientation of the muscularis externa layer. The contractility of longitudinal preparations of the rat oesophagus appeared less susceptible to the deleterious effects of acid challenge. In contrast, the responsiveness of ring-like preparations from the gastro-oesophageal junction to contractile stimulation was unaltered by acid challenge. Taurodeoxycholic acid decreased the responsiveness of circular oesophageal preparations to KCl, an effect that was exacerbated by luminal acidity. On the contrary, although the relaxant ability of the rat oesophagus did not change, acid challenge increased the relaxant efficacy of sodium nitroprusside and isoprenaline in strips of the gastro-oesophageal junction. A significant decrease in transepithelial electrical resistance was seen when the oesophageal mucosa was challenged at pH 1 but not at pH 4. Treatment with alginate blunted the deleterious effects of acid challenge on transepithelial electrical resistance and the responsiveness of oesophageal preparations to KCl. The present findings support the notion that luminal acidity is an important factor that disrupts barrier function in the oesophagus to allow the diffusion of noxious agents, such as bile acid, that alter the contractile status of the oesophagus.