MilliporeSigma
  • Characterization of Sparkling Wines According to Polyphenolic Profiles Obtained by HPLC-UV/Vis and Principal Component Analysis.
Characterization of Sparkling Wines According to Polyphenolic Profiles Obtained by HPLC-UV/Vis and Principal Component Analysis.
Foods (Basel, Switzerland) (2019-01-13)
Anaïs Izquierdo-Llopart, Javier Saurina
ABSTRACT
Cava is a sparkling wine obtained by a secondary fermentation in its own bottle. Grape skin contains several compounds, such as polyphenols, which act like natural protectors and provide flavor and color to the wines. In this paper, a previously optimized method based on reversed phase high performance liquid chromatography (HPLC) with ultraviolet/visible (UV/Vis) detection has been applied to determine polyphenols in cava wines. Compounds have been separated in a C18 core-shell column using 0.1% formic acid aqueous solution and methanol as the components of the mobile phase. Chromatograms have been recorded at 280, 310 and 370 nm to gain information on the composition of benzoic acids, hidroxycinnamic acids and flavonoids, respectively. HPLC-UV/vis data consisting of compositional profiles of relevant analytes has been exploited to characterize cava wines produced from different base wine blends using chemometrics. Other oenological variables, such as vintage, aging or malolatic fermentation, have been fixed over all the samples to avoid their influence on the description. Principal component analysis and other statistic methods have been used to extract of the underlying information, providing an excellent discrimination of samples according to grape varieties and coupages.