MilliporeSigma
  • Incidence of Antidrug Antibodies in Rheumatoid Arthritis Patients From Argentina Treated With Adalimumab, Etanercept, or Infliximab in a Real-World Setting.
Incidence of Antidrug Antibodies in Rheumatoid Arthritis Patients From Argentina Treated With Adalimumab, Etanercept, or Infliximab in a Real-World Setting.
Journal of clinical rheumatology : practical reports on rheumatic & musculoskeletal diseases (2017-12-13)
Pablo J Maid, Ricardo Xavier, Rosa M Real, Ron Pedersen, Qi Shen, Lisa Marshall, Gaston Solano, Cecilia Elena Borlenghi, Rodolfo Pardo Hidalgo
ABSTRACT
Biologic agents may induce immune responses that could impact drug action. The aims of this study were to assess antidrug antibodies (ADAs) in patients with rheumatoid arthritis (RA) from Argentina treated with etanercept, adalimumab, or infliximab at a single visit and correlate it with efficacy outcomes. In this subset analysis of a noninterventional, multinational, cross-sectional study (NCT01981473), adult patients with RA treated continuously for 6 to 24 months with etanercept, adalimumab, or infliximab were evaluated for ADAs and trough drug concentrations of 2 days or less prior to the next scheduled dose. Efficacy measurements included Disease Activity Score based on a 28-joint count-erythrocyte sedimentation rate, low disease activity, and Health Assessment Questionnaire-Disability Index. Targeted medical history of injection site/infusion reactions, serum sickness, and thromboembolic events were reported. Baseline demographics, disease characteristics, and duration of treatment of the 119 patients (etanercept: n = 54, adalimumab: n = 52, infliximab: n = 13) were similar across all groups. No etanercept-treated patient tested positive for ADAs compared with 19 (36.5%) of 52 patients and 4 (30.8%) of 13 patients treated with adalimumab and infliximab, respectively. In adalimumab- and infliximab-treated patients, ADA presence correlated negatively with trough drug levels. A greater proportion of ADA-negative patients achieved Health Assessment Questionnaire-Disability Index of 0.5 or less and had better composite efficacy measures compared with ADA-positive patients. The rate of targeted medical events reported was low. In this subset analysis, RA patients from Argentina treated with adalimumab or infliximab, but not etanercept, tested positive for ADAs. Antidrug antibody-negative patients showed a tendency toward better clinical outcomes compared with ADA-positive patients.