MilliporeSigma
  • HDAC8, A Potential Therapeutic Target, Regulates Proliferation and Differentiation of Bone Marrow Stromal Cells in Fibrous Dysplasia.
HDAC8, A Potential Therapeutic Target, Regulates Proliferation and Differentiation of Bone Marrow Stromal Cells in Fibrous Dysplasia.
Stem cells translational medicine (2018-11-15)
Tao Xiao, Yu Fu, Weiwen Zhu, Rongyao Xu, Ling Xu, Ping Zhang, Yifei Du, Jie Cheng, Hongbing Jiang
ABSTRACT
Fibrous dysplasia (FD) is a disease of postnatal skeletal stem cells caused by activating mutations of guanine nucleotide-binding protein alpha-stimulating activity polypeptide (GNAS). FD is characterized by high proliferation and osteogenesis disorder of bone marrow stromal cells (BMSCs), resulting in bone pain, deformities, and fractures. The cAMP-CREB pathway, which is activated by GNAS mutations, is known to be closely associated with the occurrence of FD. However, so far there is no available targeted therapeutic strategy for FD, as a critical issue that remains largely unknown is how this pathway is involved in FD. Our previous study revealed that histone deacetylase 8 (HDAC8) inhibited the osteogenic differentiation of BMSCs via epigenetic regulation. Here, compared with normal BMSCs, FD BMSCs exhibited significantly high proliferation and weak osteogenic capacity in response to HDAC8 upregulation and tumor protein 53 (TP53) downregulation. Moreover, inhibition of cAMP reduced HDAC8 expression, increased TP53 expression and resulted in the improvement of FD phenotype. Importantly, HDAC8 inhibition prevented cAMP-induced cell phenotype and promoted osteogenesis in nude mice that were implanted with FD BMSCs. Mechanistically, HDAC8 was identified as a transcriptional target gene of CREB1 and its transcription was directly activated by CREB1 in FD BMSCs. In summary, our study reveals that HDAC8 associates with FD phenotype and demonstrates the mechanisms regulated by cAMP-CREB1-HDAC8 pathway. These results provide insights into the molecular regulation of FD pathogenesis, and offer novel clues that small molecule inhibitors targeting HDAC8 are promising clinical treatment for FD. Stem Cells Translational Medicine 2019;8:148&14.