MilliporeSigma
  • Dietary wheat amylase trypsin inhibitors exacerbate murine allergic airway inflammation.
Dietary wheat amylase trypsin inhibitors exacerbate murine allergic airway inflammation.
European journal of nutrition (2018-03-31)
Victor F Zevallos, Verena K Raker, Joachim Maxeiner, Petra Scholtes, Kerstin Steinbrink, Detlef Schuppan
ABSTRACT
Wheat amylase trypsin inhibitors (ATI) are dietary non-gluten proteins that activate the toll-like receptor 4 on myeloid cells, promoting intestinal inflammation. We investigated the effects of dietary ATI on experimental allergic airway inflammation. Mice on a gluten and ATI-free diet (GAFD), sensitized with PBS or ovalbumin (OVA) and challenged with OVA, were compared to mice on a commercial standard chow, a gluten diet naturally containing ~ 0.75% of protein as ATI (G+AD), a gluten diet containing ~ 0.19% of protein as ATI (G-AD) and a GAFD with 1% of protein as ATI (AD). Airway hyperreactivity (AHR), inflammation in bronchoalveolar lavage (BAL) and pulmonary tissue sections were analyzed. Allergic sensitization was assessed ex vivo via proliferation of OVA-stimulated splenocytes. Mice on a GAFD sensitized with PBS did not develop AHR after local provocation with methacholine. Mice on a GAFD or on a G-AD and sensitized with OVA developed milder AHR compared to mice fed a G+AD or an AD. The increased AHR was paralleled by increased BAL eosinophils, IL-5 and IL-13 production, and an enhanced ex vivo splenocyte activation in the ATI-fed groups. Dietary ATI enhance allergic airway inflammation in OVA-challenged mice, while an ATI-free or ATI-reduced diet has a protective effect on AHR. Nutritional wheat ATI, activators of intestinal myeloid cells, may be clinically relevant adjuvants to allergic airway inflammation.