MilliporeSigma
  • Intravitreal Ets1 siRNA alleviates choroidal neovascularization in a mouse model of age-related macular degeneration.
Intravitreal Ets1 siRNA alleviates choroidal neovascularization in a mouse model of age-related macular degeneration.
Cell and tissue research (2019-03-06)
Manhui Zhu, Li Jiang, You Yuan, Lili Chen, Xiaojuan Liu, Juan Liang, Qiujian Zhu, Dongmei Ding, E Song
ABSTRACT
Choroidal neovascularization (CNV) is the basic feature of neovascular age-related macular degeneration (AMD), the leading cause of blindness in elders. Macrophages and microglia promote CNV via producing pro-angiogenic factors and inflammatory cytokines. Transcription factor E26 transformation specific-1 (Ets1) plays a pro-angiogenic role via its pro-inflammatory function. In this study, Ets1 increased and localized in the macrophages and microglia of a mouse laser-induced CNV region. Ets1 siRNA intravitreal injection ameliorated the leakage and area of CNV, as well as inhibiting the dysfunction of retinal pigment epithelium (RPE) cells and the activation of macrophages/microglia. Taken together, we provide a new insight into the molecular mechanism of CNV progression, in which Ets1 can be a new therapeutic target.