MilliporeSigma
Discovery of a novel Raf kinase inhibitor.
Endocrine-related cancer (2001-09-22)
J F Lyons, S Wilhelm, B Hibner, G Bollag
ABSTRACT
We discuss the biology of Ras signal transduction and the epidemiology of ras mutations in association with disease as a background for the development of a Raf kinase inhibitor, BAY 43-9006. Knowledge of Ras effector pathways has permitted genetic validation of numerous targets involved in the Ras signaling cascade. A key Ras effector pathway involves the kinase cascade RAF/MEK/ERK (MEK: MAP/ERK kinase; ERK: extracellular signal related kinase). Indeed, we present studies of cell lines stably expressing mutant MEK constructs, which point to Raf kinase as a target for therapeutics with selective anti-tumor activity. Finally, a small molecule drug discovery program based on inhibition of Raf kinase activity is outlined and the initial pre-clinical development process of the Raf kinase inhibitor BAY 43-9006 is discussed.
Sigma-Aldrich
Sorafenib, ≥98% (HPLC)