MilliporeSigma
Rare Streptomyces sp. polyketides as modulators of K-Ras localisation.
Organic & biomolecular chemistry (2014-05-31)
Angela A Salim, Xue Xiao, Kwang-Jin Cho, Andrew M Piggott, Ernest Lacey, John F Hancock, Robert J Capon
ABSTRACT
Chemical investigations of a soil-derived Streptomyces sp. led to the isolation of five new polyketides, (+)-oxanthromicin, (±)-hemi-oxanthromicins A/B, (±)-spiro-oxanthromicin A and oxanthroquinone, and the known alkaloid staurosporine, and the detection of four new metastable analogues, (±)-spiro-oxanthromicins B1/B2/C1/C2. Among the compounds tested, SAR investigations established that the synthetic oxanthroquinone ethyl ester and 3-O-methyl-oxanthroquinone ethyl ester were optimal at mislocalising oncogenic mutant K-Ras from the plasma membrane of intact Madin-Darby canine kidney (MDCK) cells (IC50 4.6 and 1.2 μM), while a sub-EC50 dose of (±)-spiro-oxanthromicin A was optimal at potentiating (750%) the K-Ras inhibitory activity of staurosporine (IC50 60 pM). These studies demonstrate that a rare class of Streptomyces polyketide modulates K-Ras plasma membrane localisation, with implications for the future treatment of K-Ras dependent cancers.
Sigma-Aldrich
Oxanthroquinone G01, ≥98% (HPLC)