MilliporeSigma
  • Interactions of drugs and an oligonucleotide with charged membranes analyzed by immobilized liposome chromatography.
Interactions of drugs and an oligonucleotide with charged membranes analyzed by immobilized liposome chromatography.
Biomedical chromatography : BMC (2005-06-15)
Anna Lundquist, Caroline Engvall, Elisabet Boija, Sanela Kurtovic, Jyoti Chattopadhyaya, Christine Lagerquist Hägglund, Per Lundahl
ABSTRACT
We studied the effect of charged lipids or detergent on the retention of drugs and an oligonucleotide by immobilized liposome chromatography to characterize solute-membrane interactions. This is a novel approach in analysis of oligonucleotide-liposome interactions. The charged lipids (phosphatidylserine or distearoyltrimethylammoniumpropane) or detergent (sodium dodecylsulfate) interacted electrostatically in a concentration-dependent matter with the solutes. The oligonucleotide ions presumably bound to the liposomes by multipoint interactions, which was saturable. Sodium dodecylsulfate seemed to affect the drug-membrane interactions more strongly than phosphatidylserine did, probably due to different positioning in the bilayer.
Sigma-Aldrich
Superdex®, Superdex®200 Prep Grade