MilliporeSigma
  • Highly potent, selective, and low cost bis-tetrahydroaminacrine inhibitors of acetylcholinesterase. Steps toward novel drugs for treating Alzheimer's disease.
Highly potent, selective, and low cost bis-tetrahydroaminacrine inhibitors of acetylcholinesterase. Steps toward novel drugs for treating Alzheimer's disease.
The Journal of biological chemistry (1996-09-27)
Y P Pang, P Quiram, T Jelacic, F Hong, S Brimijoin
ABSTRACT
We report highly potent, selective, and low cost bifunctional acetylcholinesterase (AChE) inhibitors developed by our two-step prototype optimization strategy utilizing computer modeling of ligand docking with target proteins: 1) identify low affinity sites normally missed by x-ray crystallography; and 2) design bifunctional analogs capable of simultaneous binding at the computer-determined low affinity site and the x-ray-identified high affinity site. Applying this strategy to 9-amino-1,2,3,4-tetrahydroacridine (THA), a drug for Alzheimer's disease, we obtained alkylene linked bis-THA analogs. These analogs were up to 10,000-fold more selective and 1,000-fold more potent than THA in inhibiting rat AChE and yet required one simple reaction to synthesize. Additionally, alkylene linked benzyl-THA analogs were developed to examine the specificity of the docking-derived low affinity THA peripheral site in AChE. The present work and our previous computational studies strongly suggest that a low affinity THA peripheral site exists in AChE. This peripheral site provides a structural basis for design of improved cholinesterase ligands for treating Alzheimer's disease and for other health-related purposes.