MilliporeSigma
  • Mutations in 16S rRNA and ribosomal protein S5 associated with high-level spectinomycin resistance in Pasteurella multocida.
Mutations in 16S rRNA and ribosomal protein S5 associated with high-level spectinomycin resistance in Pasteurella multocida.
Antimicrobial agents and chemotherapy (2007-03-21)
Corinna Kehrenberg, Stefan Schwarz
ABSTRACT
Pasteurella multocida isolates with high-level spectinomycin resistance in which no adenyltransferase genes could be demonstrated exhibited a C1192G transversion in the 16S rRNA of all six or five of the six rRNA operons and/or two different types of 3-bp deletions in the rpsE gene that codes for the ribosomal protein S5.