MilliporeSigma
  • Effects of tumor necrosis factor alpha and Interferon gamma on the viability and mRNA expression of TNF receptor type I in endothelial cells from the bovine corpus luteum
Effects of tumor necrosis factor alpha and Interferon gamma on the viability and mRNA expression of TNF receptor type I in endothelial cells from the bovine corpus luteum
Journal of Reproduction and Development (2010)
Hojo T, et al.