MilliporeSigma
  • Determinants of substrate specificity for saccharopine dehydrogenase from Saccharomyces cerevisiae.
Determinants of substrate specificity for saccharopine dehydrogenase from Saccharomyces cerevisiae.
Biochemistry (2007-06-05)
Hengyu Xu, Ann H West, Paul F Cook
ABSTRACT
A survey of NADH, alpha-Kg, and lysine analogues has been undertaken in an attempt to define the substrate specificity of saccharopine dehydrogenase and to identify functional groups on all substrates and dinucleotides important for substrate binding. A number of NAD analogues, including NADP, 3-acetylpyridine adenine dinucleotide (3-APAD), 3-pyridinealdehyde adenine dinucleotide (3-PAAD), and thionicotinamide adenine dinucleotide (thio-NAD), can serve as a substrate in the oxidative deamination reaction, as can a number of alpha-keto analogues, including glyoxylate, pyruvate, alpha-ketobutyrate, alpha-ketovalerate, alpha-ketomalonate, and alpha-ketoadipate. Inhibition studies using nucleotide analogues suggest that the majority of the binding energy of the dinucleotides comes from the AMP portion and that distinctly different conformations are generated upon binding of the oxidized and reduced dinucleotides. Addition of the 2'-phosphate as in NADPH causes poor binding of subsequent substrates but has little effect on coenzyme binding and catalysis. In addition, the 10-fold decrease in affinity of 3-APAD in comparison to NAD suggests that the nicotinamide ring binding pocket is hydrophilic. Extensive inhibition studies using aliphatic and aromatic keto acid analogues have been carried out to gain insight into the keto acid binding pocket. Data suggest that a side chain with three carbons (from the alpha-keto group up to and including the side chain carboxylate) is optimal. In addition, the distance between the C1-C2 unit and the C5 carboxylate of the alpha-keto acid is also important for binding; the alpha-oxo group contributes a factor of 10 to affinity. The keto acid binding pocket is relatively large and flexible and can accommodate the bulky aromatic ring of a pyridine dicarboxylic acid and a negative charge at the C3 but not the C4 position. However, the amino acid binding site is hydrophobic, and the optimal length of the hydrophobic portion of the amino acid carbon side chain is three or four carbons. In addition, the amino acid binding pocket can accommodate a branch at the gamma-carbon, but not at the beta-carbon.