MilliporeSigma
  • Magnetic mesoporous silica nanoparticles: fabrication and their laccase immobilization performance.
Magnetic mesoporous silica nanoparticles: fabrication and their laccase immobilization performance.
Bioresource technology (2010-07-27)
Feng Wang, Chen Guo, Liang-rong Yang, Chun-Zhao Liu
ABSTRACT
Newly large-pore magnetic mesoporous silica nanoparticles (MMSNPs) with wormhole framework structures were synthesized for the first time by using tetraethyl orthosilicate as the silica source and amine-terminated Jeffamine surfactants as template. Iminodiacerate was attached on these MMSNPs through a silane-coupling agent and chelated with Cu(2+). The Cu(2+)-chelated MMSNPs (MMSNPs-CPTS-IDA-Cu(2+)) showed higher adsorption capacity of 98.1 mg g(-1)-particles and activity recovery of 92.5% for laccase via metal affinity adsorption in comparison with MMSNPs via physical adsorption. The Michaelis constant (K(m)) and catalytic constant (k(cat)) of laccase immobilized on the MMSNPs-CPTS-IDA-Cu(2+) were 3.28 mM and 155.4 min(-1), respectively. Storage stability and temperature endurance of the immobilized laccase on MMSNPs-CPTS-IDA-Cu(2+) increased significantly, and the immobilized laccase retained 86.6% of its initial activity after 10 successive batch reactions operated with magnetic separation.
Product Description
Millipore
Amine-terminated magnetic particles, 50 mg/mL in deionized water