MilliporeSigma
  • Postsynthetic DNA modification through the copper-catalyzed azide-alkyne cycloaddition reaction.
Postsynthetic DNA modification through the copper-catalyzed azide-alkyne cycloaddition reaction.
Angewandte Chemie (International ed. in English) (2008-09-25)
Philipp M E Gramlich, Christian T Wirges, Antonio Manetto, Thomas Carell
ABSTRACT
The attachment of labels onto DNA is of utmost importance in many areas of biomedical research and is valuable in the construction of DNA-based functional nanomaterials. The copper(I)-catalyzed Huisgen cycloaddition of azides and alkynes (CuAAC) has recently been added to the repertoire of DNA labeling methods, thus allowing the virtually unlimited functionalization of both small synthetic oligonucleotides and large gene fragments with unprecedented efficiency. The CuAAC reaction yields the labeled polynucleotides in very high purity after a simple precipitation step. The reviewed technology is currently changing the way in which functionalized DNA strands are generated cost-efficiently in high quality for their application in molecular diagnostics systems and nanotechnological research.
Sigma-Aldrich
Fluorescein Azide, ≥95%
Pricing and availability is not currently available.