MilliporeSigma
  • Oligomerization of Cavbeta subunits is an essential correlate of Ca2+ channel activity.
Oligomerization of Cavbeta subunits is an essential correlate of Ca2+ channel activity.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2010-08-25)
Qi Zong Lao, Evgeny Kobrinsky, Zhuo Liu, Nikolai M Soldatov
ABSTRACT
Voltage-gated calcium channels conduct Ca(2+) ions in response to membrane depolarization. The resulting transient increase in cytoplasmic free calcium concentration is a critical trigger for the initiation of such vital responses as muscle contraction and transcription. L-type Ca(v)1.2 calcium channels are complexes of the pore-forming α(1C) subunit associated with cytosolic Ca(v)β subunits. All major Ca(v)βs share a highly homologous membrane associated guanylate kinase-like (MAGUK) domain that binds to α(1C) at the α-interaction domain (AID), a short motif in the linker between transmembrane repeats I and II. In this study we show that Ca(v)β subunits form multimolecular homo- and heterooligomeric complexes in human vascular smooth muscle cells expressing native calcium channels and in Cos7 cells expressing recombinant Ca(v)1.2 channel subunits. Ca(v)βs oligomerize at the α(1C) subunits residing in the plasma membrane and bind to the AID. However, Ca(v)β oligomerization occurs independently on the association with α(1C). Molecular structures responsible for Ca(v)β oligomerization reside in 3 regions of the guanylate kinase subdomain of MAGUK. An augmentation of Ca(v)β homooligomerization significantly increases the calcium current density, while heterooligomerization may also change the voltage-dependence and inactivation kinetics of the channel. Thus, oligomerization of Ca(v)β subunits represents a novel and essential aspect of calcium channel regulation.
Sigma-Aldrich
Gel Filtration Markers Kit for Protein Molecular Weights 29,000-700,000 Da