MilliporeSigma
  • Transport of Nucleoside Triphosphates into Cells by Artificial Molecular Transporters.
Transport of Nucleoside Triphosphates into Cells by Artificial Molecular Transporters.
Angewandte Chemie (International ed. in English) (2018-03-27)
Zbigniew Zawada, Ameneh Tatar, Pavle Mocilac, Miloš Buděšínský, Tomáš Kraus
ABSTRACT
Chemically modified nucleoside triphosphates (NTPs) are widely exploited as unnatural metabolites in chemical biology and medicinal chemistry. Because anionic NTPs do not permeate cell membranes, their corresponding neutral precursors are employed in cell-based assays. These precursors become active metabolites after enzymatic conversion, which often proceeds insufficiently. Here we show that metabolically-active NTPs can be directly transported into eukaryotic cells and bacteria by the action of designed synthetic nucleoside triphosphate transporters (SNTTs). The transporter is composed of a receptor, which forms a non-covalent complex with a triphosphate anion, and a cell-penetrating agent, which translocates the complex across the plasma membrane. NTP is then released from the complex in the intracellular milieu and accumulates in nuclei and nucleoli in high concentration. The transport of NTPs proceeds rapidly (seconds to minutes) and selectively even in the presence of other organic anions. We demonstrate that this operationally simple and efficient means of transport of fluorescently labelled NTPs into cells can be used for metabolic labeling of DNA in live cells.
Sigma-Aldrich
BioTracker NTP-Transporter Molecule, Quick and efficient transport of labeled NTPs into cells for live cell imaging of DNA replication, synthesis, etc. Labeling of DNA with fluorescent NTPs is one of the essential methods in cell cycle analysis.
Pricing and availability is not currently available.