MilliporeSigma
  • Notch4 and Wnt-1 proteins function to regulate branching morphogenesis of mammary epithelial cells in an opposing fashion.
Notch4 and Wnt-1 proteins function to regulate branching morphogenesis of mammary epithelial cells in an opposing fashion.
Developmental biology (1998-05-13)
H Uyttendaele, J V Soriano, R Montesano, J Kitajewski
ABSTRACT
Elongation and branching of epithelial ducts is a crucial event during the development of the mammary gland. Branching morphogenesis of the mouse mammary epithelial TAC-2 cell line was used as an assay to examine the role of Wnt, HGF, TGF-beta, and the Notch receptors in branching morphogenesis. Wnt-1 was found to induce the elongation and branching of epithelial tubules, like HGF and TGF-beta 2, and to strongly cooperate with either HGF or TGF-beta 2 in this activity. Wnt-1 displayed morphogenetic activity in TAC-2 cells as it induced branching even under conditions that normally promote cyst formation. The Notch4(int-3) mammary oncoprotein, an activated form of the Notch4 receptor, inhibited the branching morphogenesis normally induced by HGF and TGF-beta 2. The minimal domain within the Notch4(int-3) protein required to inhibit morphogenesis consists of the CBF-1 interaction domain and the cdc10 repeat domain. Coexpression of Wnt-1 and Notch4(int-3) demonstrates that Wnt-1 can overcome the Notch-mediated inhibition of branching morphogenesis. These data suggest that Wnt and Notch signaling may play opposite roles in mammary gland development, a finding consistent with the convergence of the wingless and Notch signaling pathways found in Drosophila.
Sigma-Aldrich
Collagen Type I, rat tail