Merck
  • Standardised herbal extract of chlorogenic acid from leaves of Etlingera elatior (Zingiberaceae).
Standardised herbal extract of chlorogenic acid from leaves of Etlingera elatior (Zingiberaceae).
Pharmacognosy research (2011-10-25)
Eric W C Chan, Yau Yan Lim, Shiau Pin Tan
ABSTRACT
Chlorogenic acid (CGA) or 5-caffeoylquinic acid, was found to be the dominant phenolic compound in leaves of Etlingera elatior (Zingiberaceae). The CGA content of E. elatior leaves was significantly higher than flowers of Lonicera japonica (honeysuckle), the commercial source. In this study, a protocol to produce a standardised herbal CGA extract from leaves of E. elatior using column chromatography was developed. Freeze-dried leaves of E. elatior were extracted with 30% ethanol, and sequentially fractionated using Diaion HP-20 and Sephadex LH-20. The CGA fractions, which yielded extracts of 10% and 40% w/w purity, possessed antioxidant, tyrosinase inhibition, and antibacterial properties. The entire fractionation process took only 6.5 hours, using gravity flow. From 50 g of leaves, the final yield of CGA extract was 0.2 g (0.4%). The CGA content of the standardised herbal extract from leaves of E. elatior (40%) is 1.6 times that of commercial extracts from honeysuckle flowers (25%). With high CGA content, the standardised herbal extract has a great potential to be developed into functional food and other health products. Leaves of E. elatior, which currently have no economic value, could serve as an alternative source of CGA. Leaves are large, available in abundance, and harvesting is non-destructive to the plants.
Product Description
Supelco
Sephadex® LH-20
Pricing and availability is not currently available.
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Pricing and availability is not currently available.
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Pricing and availability is not currently available.