MilliporeSigma
  • Casein kinase 1alpha governs antigen-receptor-induced NF-kappaB activation and human lymphoma cell survival.
Casein kinase 1alpha governs antigen-receptor-induced NF-kappaB activation and human lymphoma cell survival.
Nature (2009-01-02)
Nicolas Bidère, Vu N Ngo, Jeansun Lee, Cailin Collins, Lixin Zheng, Fengyi Wan, R Eric Davis, Georg Lenz, D Eric Anderson, Damien Arnoult, Aimé Vazquez, Keiko Sakai, Jun Zhang, Zhaojing Meng, Timothy D Veenstra, Louis M Staudt, Michael J Lenardo
ABSTRACT
The transcription factor NF-kappaB is required for lymphocyte activation and proliferation as well as the survival of certain lymphoma types. Antigen receptor stimulation assembles an NF-kappaB activating platform containing the scaffold protein CARMA1 (also called CARD11), the adaptor BCL10 and the paracaspase MALT1 (the CBM complex), linked to the inhibitor of NF-kappaB kinase complex, but signal transduction is not fully understood. We conducted parallel screens involving a mass spectrometry analysis of CARMA1 binding partners and an RNA interference screen for growth inhibition of the CBM-dependent 'activated B-cell-like' (ABC) subtype of diffuse large B-cell lymphoma (DLBCL). Here we report that both screens identified casein kinase 1alpha (CK1alpha) as a bifunctional regulator of NF-kappaB. CK1alpha dynamically associates with the CBM complex on T-cell-receptor (TCR) engagement to participate in cytokine production and lymphocyte proliferation. However, CK1alpha kinase activity has a contrasting role by subsequently promoting the phosphorylation and inactivation of CARMA1. CK1alpha has thus a dual 'gating' function which first promotes and then terminates receptor-induced NF-kappaB. ABC DLBCL cells required CK1alpha for constitutive NF-kappaB activity, indicating that CK1alpha functions as a conditionally essential malignancy gene-a member of a new class of potential cancer therapeutic targets.
Product Description
Millipore
Immobilon Western Chemiluminescent HRP Substrate, Immobilon Western HRP Substrate provides high sensitivity for Chemiluminescent detection in western or dot/slot/spot blotting applications on both PVDF and nitrocellulose membranes, and is compatible with all commonly used buffers and blocking reagents.